Learn More
Bio Rad Laboratories AG MP-1M RESIN 500G
Catalog No. NC1411275
Encompass
500 g, analytical grade anion exchange resin, chloride form, macroporous, 100–200 dry mesh size, 75–150 µm wet bead size, >1,000,000 MW limit
Catalog No. NC1411275 Supplier Bio Rad Laboratories Supplier No. 1411841
500 g, analytical grade anion exchange resin, chloride form, macroporous, 100–200 dry mesh size, 75–150 µm wet bead size, >1,000,000 MW limit