Learn More
Nanocomposix BioPure Silver Nanospheres - Bare (Citrate) - 20 nm, 1 mg/mL in 2 mM sodium citrate, 5 mL
SDP
Catalog No. 501056961
Encompass_Preferred
As a global leader in silver nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.20 nm spherical silver nanoparticles combine strong plasmonic optical properties with antimicrobial capabilities. These nanoparticles are useful in a broad range of applications including conductive composites, biosensors, sensing, photovoltaics, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. 50-105-6961 Supplier Nanocomposix Supplier No. AGCB205M
As a global leader in silver nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.20 nm spherical silver nanoparticles combine strong plasmonic optical properties with antimicrobial capabilities. These nanoparticles are useful in a broad range of applications including conductive composites, biosensors, sensing, photovoltaics, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.