Learn More
Enzo Life Sciences Antigen Retrieval Reagent, pH 9 (10X) (100ml)
SDP
Catalog No. 502013361
Encompass_Preferred
Antigen Retrieval Reagent, pH 9 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin. Formulation: Liquid. In Tris-EDTA buffer. Long Term Storage: +4°C. Applications: IHC .
Catalog No. 50-201-3361 Supplier Enzo Life Sciences Supplier No. ENZACC1130100
Antigen Retrieval Reagent, pH 9 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin. Formulation: Liquid. In Tris-EDTA buffer. Long Term Storage: +4°C. Applications: IHC .