Learn More
Bio Rad Laboratories AG1-X4 ANIONEXCHANGERESIN 500G
Catalog No. NC2290730
Encompass
AG 1-X4 Anion Exchange Resin, analytical grade, 100–200 mesh, chloride form, 500 g #1401341500 g, analytical grade anion exchange resin, chloride form, 4% crosslinkage, 100–200 dry mesh size, 106–250 µm wet bead size, ~1,400 MW limit
This item is not returnable. View return policy
Catalog No. NC2290730 Supplier Bio Rad Laboratories Supplier No. 1401341
AG 1-X4 Anion Exchange Resin, analytical grade, 100–200 mesh, chloride form, 500 g #1401341500 g, analytical grade anion exchange resin, chloride form, 4% crosslinkage, 100–200 dry mesh size, 106–250 µm wet bead size, ~1,400 MW limit