Learn More
Medchemexpress LLC HY-10253 10mg Medchemexpress, AG1024 CAS:65678-07-1 Purity:>98%
SDP
Catalog No. 501967991
Encompass_Preferred
Medchemexpress, HY-10253 10mg AG1024 CAS:65678-07-1 AG1024 (Tyrphostin AG 1024) is a reversible, competitive and selective insulin-like growth factor-1 (IGF-1) inhibitor with an IC50 of 7 μM for phosphorylation of IGF1R. AG1024 inhibits phosphorylation of insulin receptor (IC50=57 μM). AG1024 induces apoptosis and has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7991 Supplier Medchemexpress LLC Supplier No. HY1025310MG
Medchemexpress, HY-10253 10mg AG1024 CAS:65678-07-1 AG1024 (Tyrphostin AG 1024) is a reversible, competitive and selective insulin-like growth factor-1 (IGF-1) inhibitor with an IC50 of 7 μM for phosphorylation of IGF1R. AG1024 inhibits phosphorylation of insulin receptor (IC50=57 μM). AG1024 induces apoptosis and has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.