Learn More
Medchemexpress LLC HY-108330 5mg Medchemexpress, AG126 CAS:118409-62-4 Purity:>98%
SDP
Catalog No. 501968531
Encompass_Preferred
Medchemexpress, HY-108330 5mg AG126 CAS:118409-62-4 AG126 is a tyrosine kinase inhibitor which can prevent the activation of mitogen-activated protein kinase p42MAPK (ERK2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8531 Supplier Medchemexpress LLC Supplier No. HY1083305MG
Medchemexpress, HY-108330 5mg AG126 CAS:118409-62-4 AG126 is a tyrosine kinase inhibitor which can prevent the activation of mitogen-activated protein kinase p42MAPK (ERK2). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.