Learn More
Medchemexpress LLC HY-12032 5mg Medchemexpress, AG14361 CAS:328543-09-5 Purity:>98%
SDP
Catalog No. 501969600
Encompass_Preferred
Medchemexpress, HY-12032 5mg AG14361 CAS:328543-09-5 AG14361 is a potent PARP-1 inhibitor, with a Ki of < 5 nM, and in permeabilized SW620 and intact SW620 cells, the IC50s are 29 nM and 14 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9600 Supplier Medchemexpress LLC Supplier No. HY120325MG
Medchemexpress, HY-12032 5mg AG14361 CAS:328543-09-5 AG14361 is a potent PARP-1 inhibitor, with a Ki of < 5 nM, and in permeabilized SW620 and intact SW620 cells, the IC50s are 29 nM and 14 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.