Learn More
Thermo Scientific AG14A-7 MICRON GUARD COLUMN
Catalog No. NC9314211 Shop All Thermo Scientific Products
Encompass
IonPac AG14A-7 um Guard Column (4 x 50 mm)
Catalog No. NC9314211 Supplier Thermo Scientific Supplier No. 056897
IonPac AG14A-7 um Guard Column (4 x 50 mm)