Learn More
TARGETMOL CHEMICALS INC AG490 200MG
SDP
Catalog No. 502877537
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AG490 (Tyrphostin B42) is an inhibitor of EGFR (IC50 0.1 uM). It is 135-fold more selective for EGFR than ErbB2, also inhibits JAK2 with no effect to Lyn, Lck, Syk, Btk, and Src. Purity 99.7%
Catalog No. 50-287-7537 Supplier TARGETMOL CHEMICALS INC Supplier No. T2600200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. AG490 (Tyrphostin B42) is an inhibitor of EGFR (IC50 0.1 uM). It is 135-fold more selective for EGFR than ErbB2, also inhibits JAK2 with no effect to Lyn, Lck, Syk, Btk, and Src. Purity 99.7%