Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP7382EC10UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: E. coli. Species Reactivity: M. tuberculosis. Protein Expression: A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Ag85C, was expressed in the hosts and tags indicated. AA Sequence: AFSRPGLPVE YLQVPSASMG RDIKVQFQGG GPHAVYLLDG LRAQDDYNGW DINTPAFEEY YQSGLSVIMP VGGQSSFYTD WYQPSQSNGQ NYTYKWETFL TREMPAWLQA NKGVSPTGNA AVGLSMSGGS ALILAAYYPQ QFPYAASLSG FLNPSEGWWP TLIGLAMNDS GGYNANSMWG PSSDPAWKRN DPMVQIPRLV ANNTRIWVYC GNGTPSDLGG DNIPAKFLEG LTLRTNQTFR DTYAADGGRN GVFNFPPNGT HSWPYWNEQL VAMKADIQHV LNGATPPAAP AAPAA. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: fbpC. Uniprot: P9WQN8.
Catalog No. 50-188-7036