Learn More
Aviva Systems Biology Corp AGA Antibody - middle region (ARP61039_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61039_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGA. It was validated on Western Blot. . Alternate names: AGU, ASRG, GAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-287-238