Learn More
Cayman Chemical aGalactosIdase A RabbIt Mon
SDP
Catalog No. 502916344
Encompass_Preferred
An imidazole displays excitation/emission maxima of 382/466 nm respectively when complexed with trichloroacetic acid
Catalog No. 50-291-6344 Supplier Cayman Chemical Supplier No. 38088100 UL
An imidazole displays excitation/emission maxima of 382/466 nm respectively when complexed with trichloroacetic acid