Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aganepag | 910562-18-4 | 99.7% | 429.57 | C24H31NO4S | 10 MG
SDP

Catalog No. 5000296781
Encompass_Preferred

Aganepag is a potent prostanoid EP2 receptor agonist for research use, applied in preclinical studies of EP2 signaling, wound healing, scar reduction, and dermatological research. It has a low EC50 and defined chemical properties suitable for biochemical and cell-based assays.

  • Potent EP2 receptor agonist (EC50 0.19 nM).
  • High purity (~99.7%).
  • Molecular weight 429.57.
  • Chemical formula C24H31NO4S.
  • Suitable for in vitro and preclinical research applications.
  • Available in small research pack sizes such as 10 MG.

Catalog No. 50-002-96781 Supplier Medchemexpress LLC Supplier No. HY1986410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aganepag is a potent prostanoid EP2 receptor agonist for research use, applied in preclinical studies of EP2 signaling, wound healing, scar reduction, and dermatological research. It has a low EC50 and defined chemical properties suitable for biochemical and cell-based assays.

  • Potent EP2 receptor agonist (EC50 0.19 nM).
  • High purity (~99.7%).
  • Molecular weight 429.57.
  • Chemical formula C24H31NO4S.
  • Suitable for in vitro and preclinical research applications.
  • Available in small research pack sizes such as 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.