Learn More
Alkali Scientific Agar, 1 Gram Capsules, High Gel Strength, 1 Kilogram
SDP
Catalog No. 5000660156
Encompass_Preferred
High gel strength phyto-grade agar is available in capsule form to provide a more convenient and consistent option for plant tissue culture applications. The agar is pre-measured into 1-gram capsules, allowing for easy preparation without the need for weighing or measuring. This agar provides the necessary solid support for plant growth while maintaining optimal pH and gel strength during incubation. The 1kg package offers an economical option for larger-scale plant culture operations, and the capsules ensure uniformity in each batch, reducing preparation time and minimizing potential errors in agar preparation for plant research and biotechnology projects.
Catalog No. 50-006-60156 Supplier Alkali Scientific Supplier No. AGB004CP
High gel strength phyto-grade agar is available in capsule form to provide a more convenient and consistent option for plant tissue culture applications. The agar is pre-measured into 1-gram capsules, allowing for easy preparation without the need for weighing or measuring. This agar provides the necessary solid support for plant growth while maintaining optimal pH and gel strength during incubation. The 1kg package offers an economical option for larger-scale plant culture operations, and the capsules ensure uniformity in each batch, reducing preparation time and minimizing potential errors in agar preparation for plant research and biotechnology projects.