Learn More
Research Products International Corp Agar, 1 Gram Capsules, High Gel Strength, 500 Grams
SDP
Catalog No. 50488453
Encompass_Preferred
Each capsule contains 1g Agar. Convenient and easy to use, simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.
Catalog No. 50-488-453 Supplier Research Products International Corp Supplier No. A20021500.0
Each capsule contains 1g Agar. Convenient and easy to use, simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.