Learn More
LAMDA BIOTECH INC Agar
Catalog No. 502045829
Encompass_Preferred
High quality agar, derived from Gelidium seaweed. This product is widely used for general bacteriological research for making various microbiological media and other tissue culture media.
Catalog No. 50-204-5829 Supplier LAMDA BIOTECH INC Supplier No. C110
High quality agar, derived from Gelidium seaweed. This product is widely used for general bacteriological research for making various microbiological media and other tissue culture media.