Learn More
Crescent Chemical Co Inc AGAR AGAR SERVA HIGH GEL
SDP
Catalog No. 50247206
Encompass_Preferred
Agar Agar SERVA high gel-strength powder research grade Choice quality for in vitro culture of plants or bacteriology, no turbidity with phosphates. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11396.04 CAS 9002-18-0
Catalog No. 50-247-206 Supplier Crescent Chemical Co Inc Supplier No. SE11396.04
Agar Agar SERVA high gel-strength powder research grade Choice quality for in vitro culture of plants or bacteriology, no turbidity with phosphates. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11396.04 CAS 9002-18-0