Learn More
Crescent Chemical Co Inc AGAR AGAR SERVA HIGH GEL
SDP
Supplier:  Crescent Chemical Co Inc SE11396.03
Encompass_Preferred
Agar Agar SERVA high gel-strength powder research grade Choice quality for in vitro culture of plants or bacteriology, no turbidity with phosphates. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11396.03 CAS 9002-18-0
Catalog No. 50-247-205