Learn More
Crescent Chemical Co Inc AGAR AGAR SERVA KOBE I
SDP
Catalog No. 50247203
Encompass_Preferred
Agar Agar SERVA Kobe I research grade, in stripes Threadlike, bleached; tested for use in nutrient media. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11395.04 CAS 9002-18-0
Catalog No. 50-247-203 Supplier Crescent Chemical Co Inc Supplier No. SE11395.04
Agar Agar SERVA Kobe I research grade, in stripes Threadlike, bleached; tested for use in nutrient media. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11395.04 CAS 9002-18-0