Learn More
Crescent Chemical Co Inc AGAR AGAR SERVA POWDER
SDP
Supplier:  Crescent Chemical Co Inc SE11393.03
Encompass_Preferred
Agar Agar SERVA powder analytical grade Highly purified. Suitable for immunodiffusion. Size - 250G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11393.03 CAS 9002-18-0
Catalog No. 50-247-200