Learn More
Crescent Chemical Co Inc AGAR AGAR SERVA POWDER
SDP
Catalog No. 50247201
Encompass_Preferred
Agar Agar SERVA powder analytical grade Highly purified. Suitable for immunodiffusion. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11393.04 CAS 9002-18-0
Catalog No. 50-247-201 Supplier Crescent Chemical Co Inc Supplier No. SE11393.04
Agar Agar SERVA powder analytical grade Highly purified. Suitable for immunodiffusion. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 11393.04 CAS 9002-18-0