Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Agar Art: Creating Masterpieces with Microbes
SDPSCI_ED

Catalog No. S05962 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05962 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05962 Supplier EDVOTEK™ Supplier No. 228
Add to Cart
Add to Cart

In this 'Agar Art' experiment, students create 'Bio-Paints' using bacterial culture techniques. The color-producing microbes are painted onto an agar plate canvas. After incubation will inspect, admire, and hopefully share your living art.

  • Create works of art using color-producing microbes
  • Explore the intersections of science and art and the field of BioArt
  • Understand the transformation technologies used to create bacterial BioPaints
  • Includes four brightly colored BioPaints and the material needed for 25 agar canvases
  • Group Size: For 25 Students
  • Time Required: 60 minutes (plus two incubations)
  • Kit Includes: Instructions, ampicillin, IPTG, GFP, Pink, Blue, and Purple BactoBeads™ ReadyPour™ Luria Broth agar, sterile, Luria Broth liquid media, inoculating loops, sterile, plastic loops, non-sterile, toothpicks, petri plates, 1.5 mL microcentrifuge tubes, and 10 mL pipet, sterile
  • All You Need: Distilled water, gloves, UV light source, microwave, incubator or water bath, adjustable pipette and tips (20-200μl), pipet pump (optional), vortexer (optional), and fine tip paint brushes (optional).
  • Storage: Some Components Require Refrigerator and Freezer Storage Upon Receipt

Specifications

Grade 9 to Undergraduate
Class Size 25 Students
Product Type Transformation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.