Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar ash 2.0-4.5500G
Catalog No. 502898477
Encompass
Agar is a polysaccharide consisting of - D-galactopyranosyl and 3 6-anhydro -L- galactopyranosyl units. Depending on the source agar exists as various substituted forms such as methyl and sulphate ether. It is a linear galactan polysaccharide isolated from the cell wall of Rhodophyceae class of seaweeds.
Catalog No. 50-289-8477 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A7002500G
Agar is a polysaccharide consisting of - D-galactopyranosyl and 3 6-anhydro -L- galactopyranosyl units. Depending on the source agar exists as various substituted forms such as methyl and sulphate ether. It is a linear galactan polysaccharide isolated from the cell wall of Rhodophyceae class of seaweeds.