Learn More
Research Products International Corp Agar, Bacteriological Grade, 10 KG
SDP
Catalog No. 501976905
Encompass_Preferred
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
Catalog No. 50-197-6905 Supplier Research Products International Corp Supplier No. A2003010000.0
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.