Learn More
NEOGEN Agar No. 2 Bacteriological, 500 g
SDP
Catalog No. 502015819
Encompass_Preferred
Agar No. 2 Bacteriological is a clear bacteriological agar with high gel strength compatible with all culture media. Agar No. 2 Bacteriological is not intended for use in the diagnosis of disease or other conditions in humans.A bacteriological agar which gives a firm gel at working concentrations of 1.0 to 1.5% which is reasonably clear. This agar is recommended for all culture media except sensitivity testing media and those where absolute clarity is advantageous.For more information visit https://www.neogen.com/solutions/Microbiology/agar-number-2-bacteriological
Catalog No. 50-201-5819 Supplier NEOGEN Supplier No. 700003593
Agar No. 2 Bacteriological is a clear bacteriological agar with high gel strength compatible with all culture media. Agar No. 2 Bacteriological is not intended for use in the diagnosis of disease or other conditions in humans.A bacteriological agar which gives a firm gel at working concentrations of 1.0 to 1.5% which is reasonably clear. This agar is recommended for all culture media except sensitivity testing media and those where absolute clarity is advantageous.For more information visit https://www.neogen.com/solutions/Microbiology/agar-number-2-bacteriological