Learn More
Bioworld Agar, Bacteriological, 9002-18-0, 500g
Catalog No. 50254260
Encompass_Preferred
AgarFine, light-colored granules, Bacteriological Grade, Gel Strength: 550–950 g/cm2, Gelling Temperature: 34–38 °C, Melting Temperature: 84–88 °C, pH: 6.0–7.5, Soluble in boiling water, Appearance Form : Powder, Appearance Color : Off-white. 500g
Catalog No. 50-254-260 Supplier Bioworld Supplier No. 306200011
AgarFine, light-colored granules, Bacteriological Grade, Gel Strength: 550–950 g/cm2, Gelling Temperature: 34–38 °C, Melting Temperature: 84–88 °C, pH: 6.0–7.5, Soluble in boiling water, Appearance Form : Powder, Appearance Color : Off-white. 500g