Learn More
LabScientific Agar, Bacteriological | 1kg, Purest form of general purpose agar available. Drosophila. Pharmaceutical Grade Agar.
Catalog No. NC0261501
Encompass
Agar, bacteriological grade, 1kg. Supports the growth of a wide variety of both fastidious and non-fastidious organisms. Purest general purpose agar available. Pharmaceutical Grade Agar is manufactured using a freeze-thaw extraction process, making it one of the purest forms of general purpose agar available. 1kg.
Catalog No. NC0261501 Supplier LabScientific Supplier No. A4661KG
Agar, bacteriological grade, 1kg. Supports the growth of a wide variety of both fastidious and non-fastidious organisms. Purest general purpose agar available. Pharmaceutical Grade Agar is manufactured using a freeze-thaw extraction process, making it one of the purest forms of general purpose agar available. 1kg.