Learn More
LabScientific Pharmaceutical Grade Agar is manufactured using a freeze-thaw extraction process, making it one of the purest forms of general purpose agar available. Bacteriological, drosophila media. 2kg
Supplier:  LabScientific A4662KG
Encompass
Agar, bacteriological grade, 1kg. Supports the growth of a wide variety of both fastidious and non-fastidious organisms. Purest general purpose agar available. Pharmaceutical Grade Agar is manufactured using a freeze-thaw extraction process, making it one of the purest forms of general purpose agar available. 2kg.
Catalog No. NC0616758