Learn More
Research Products International Corp Agar, Bacteriological Grade, 5 Kilograms
SDP
Catalog No. 501368024
Encompass_Preferred
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
Catalog No. 50-136-8024 Supplier Research Products International Corp Supplier No. A200305000.0
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.