Learn More
Bioworld Agar, Bacteriological, 9002-18-0, 1kg
Catalog No. 50254261
Encompass_Preferred
AgarFine, light-colored granules, Bacteriological Grade, Gel Strength: 550–950 g/cm2, Gelling Temperature: 34–38 °C, Melting Temperature: 84–88 °C, pH: 6.0–7.5, Soluble in boiling water, Appearance Form : Powder, Appearance Color : Off-white. 1kg
Catalog No. 50-254-261 Supplier Bioworld Supplier No. 306200012
AgarFine, light-colored granules, Bacteriological Grade, Gel Strength: 550–950 g/cm2, Gelling Temperature: 34–38 °C, Melting Temperature: 84–88 °C, pH: 6.0–7.5, Soluble in boiling water, Appearance Form : Powder, Appearance Color : Off-white. 1kg