Learn More
Bioworld Bacteriological Agar, 2.5 kg
Catalog No. 50254262
Encompass_Preferred
AgarFine, light-colored granulesBacteriological GradeGel Strength: 550–950 g/cm2Gelling Temperature: 34–38 °C Melting Temperature: 84–88 °C pH: 6.0–7.5Soluble in boiling water
Catalog No. 50-254-262 Supplier Bioworld Supplier No. 306200013
AgarFine, light-colored granulesBacteriological GradeGel Strength: 550–950 g/cm2Gelling Temperature: 34–38 °C Melting Temperature: 84–88 °C pH: 6.0–7.5Soluble in boiling water