Learn More
NEOGEN Agar No. 1 Bacteriological, 10 kg
SDP
Catalog No. 502015815
Encompass_Preferred
Agar No. 1 Bacteriological is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.A high clarity agar with good gelling properties and a low concentration of metal ions. This agar is suitable for all bacteriological purposes including sensitivity testing and pour plate techniques. A firm gel is obtained at working concentrations of 1.0 to 1.5%. No significant precipitation is observed on reheating or prolonged holding at 65°C.For more information visit https://www.neogen.com/solutions/Microbiology/agar-number-1-bacteriological
Catalog No. 50-201-5815 Supplier NEOGEN Supplier No. 700004677
Agar No. 1 Bacteriological is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.A high clarity agar with good gelling properties and a low concentration of metal ions. This agar is suitable for all bacteriological purposes including sensitivity testing and pour plate techniques. A firm gel is obtained at working concentrations of 1.0 to 1.5%. No significant precipitation is observed on reheating or prolonged holding at 65°C.For more information visit https://www.neogen.com/solutions/Microbiology/agar-number-1-bacteriological