Learn More
LabScientific Agar, Bacteriological | 1kg, Purest form of general purpose agar available. Drosophila. Pharmaceutical Grade Agar.
Supplier:  LabScientific A466_BR01
Encompass
Agar, bacteriological grade, 1kg. Supports the growth of a wide variety of both fastidious and non-fastidious organisms. Purest general purpose agar available. Pharmaceutical Grade Agar is manufactured using a freeze-thaw extraction process, making it one of the purest forms of general purpose agar available. 1kg.
Catalog No. NC2067500