Learn More
Bellco Glass, Inc. Agar Bottle Complete, 80mL
SDP
Catalog No. 501945510
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass.. MADE IN THE USASpecifications: Capacity: 80ml Dimensions: 55mm wide x 135mm long x 30mm deep Dam in bottle,, 25mm from bottom Neck size; 25mm Supplied with 25mm screw cap(2535-00025)& stopper (2048-11800)
Catalog No. 50-194-5510 Supplier Bellco Glass, Inc. Supplier No. 253550020
Manufactured from type 1 class a Borosilicate glass.. MADE IN THE USASpecifications: Capacity: 80ml Dimensions: 55mm wide x 135mm long x 30mm deep Dam in bottle,, 25mm from bottom Neck size; 25mm Supplied with 25mm screw cap(2535-00025)& stopper (2048-11800)