Learn More
Research Products International Corp Agar, 1 Gram Capsules, High Gel Strength, 1 Kilogram
SDP
Supplier:  Research Products International Corp A200211000.0
Encompass_Preferred
Each capsule contains 1g Agar. Convenient and easy to use, simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.
Catalog No. 50-136-8017