Learn More
Research Products International Corp Agar, Daishin, 1 Kilogram
SDP
Catalog No. 50488476
Encompass_Preferred
Well known agar brand in plant tissue culture. Tested for micropropagation in numerous plants.
Gel Strength: 700-800 g/cm2.
Catalog No. 50-488-476 Supplier Research Products International Corp Supplier No. A203501000.0
Well known agar brand in plant tissue culture. Tested for micropropagation in numerous plants.
Gel Strength: 700-800 g/cm2.