Learn More
Bioworld Agar, Daishin, 500 g
Catalog No. 50255201
Encompass_Preferred
A substrate for inclusion in culture media, gel preparation, and microbiological work. Daishin Agar is a high quality agar used for plant culture, with good growth characteristics reported for numerous plants and tissues. Studies have shown Daishin Agar to yield highly consistent results across batches in comparison to other agars.Tested for micropropagation in numerous plantsGel Strength: 700−800 g/cm2This product is sold for laboratory research use only.Appearance: SolidHS Code : 130231000Melting Point: 85-95 °CRTECS Number: AW7950000
Catalog No. 50-255-201 Supplier Bioworld Supplier No. 401000561
A substrate for inclusion in culture media, gel preparation, and microbiological work. Daishin Agar is a high quality agar used for plant culture, with good growth characteristics reported for numerous plants and tissues. Studies have shown Daishin Agar to yield highly consistent results across batches in comparison to other agars.Tested for micropropagation in numerous plantsGel Strength: 700−800 g/cm2This product is sold for laboratory research use only.Appearance: SolidHS Code : 130231000Melting Point: 85-95 °CRTECS Number: AW7950000