Learn More
Research Products International Corp Agar, Granulated, High Gel Strength, 1 Kilogram
SDP
Catalog No. 50488473
Encompass_Preferred
Purified agar with excellent properties for tissue culture and microbiological work.
To prepare media, add 15 grams to 1 liter of water.
Catalog No. 50-488-473 Supplier Research Products International Corp Supplier No. A202501000.0
Purified agar with excellent properties for tissue culture and microbiological work.
To prepare media, add 15 grams to 1 liter of water.