Learn More
Research Products International Corp Agar, Granulated, 2 KG
SDP
Supplier:  Research Products International Corp A202502000.0
Encompass_Preferred
Purified agar with excellent properties for tissue culture and microbiological work.
To prepare media, add 15 grams to 1 liter of water.
Catalog No. 50-223-0469