Learn More
Research Products International Corp Agar, Granulated, High Gel Strength, 500 Grams
SDP
Catalog No. 50488474
Encompass_Preferred
Purified agar with excellent properties for tissue culture and microbiological work.
To prepare media, add 15 grams to 1 liter of water.
Catalog No. 50-488-474 Supplier Research Products International Corp Supplier No. A20250500.0
Purified agar with excellent properties for tissue culture and microbiological work.
To prepare media, add 15 grams to 1 liter of water.