Learn More
Bioworld Agar, Granulated, 5 kg
Catalog No. 50255204
Encompass_Preferred
A substrate for inclusion in culture media, gel preparation, and microbiological work.This product is sold for laboratory research use only.Appearance: SolidHS Code : 130231000Melting Point: 85-95 °CRTECS Number: AW7950000
Catalog No. 50-255-204 Supplier Bioworld Supplier No. 401000603
A substrate for inclusion in culture media, gel preparation, and microbiological work.This product is sold for laboratory research use only.Appearance: SolidHS Code : 130231000Melting Point: 85-95 °CRTECS Number: AW7950000