Learn More
Bioworld Agar, High Gel Strength, 500 g
Catalog No. 50255205
Encompass_Preferred
AgarPurified high gel strength agar with excellent properties for tissue culture and microbiological workGel Strength: 900 g/cm2Working Concentration: 5.0 g/LSulfated ash: <6%This product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000
Catalog No. 50-255-205 Supplier Bioworld Supplier No. 401000641
AgarPurified high gel strength agar with excellent properties for tissue culture and microbiological workGel Strength: 900 g/cm2Working Concentration: 5.0 g/LSulfated ash: <6%This product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000