Learn More
Research Products International Corp Agar, High Gel Strength, Powder, 1 Kilogram
SDP
Catalog No. 50213116
Encompass_Preferred
Purified high gel strength agar with excellent properties for tissue culture and microbiological work.
Molecular Biology Grade
Catalog No. 50-213-116 Supplier Research Products International Corp Supplier No. A200201000.0
Purified high gel strength agar with excellent properties for tissue culture and microbiological work.
Molecular Biology Grade