Learn More
Research Products International Corp Agar, High Gel Strength, Powder, 500 Grams
SDP
Catalog No. 50488450
Encompass_Preferred
Purified high gel strength agar with excellent properties for tissue culture and microbiological work.
Molecular Biology Grade
Catalog No. 50-488-450 Supplier Research Products International Corp Supplier No. A20020500.0
Purified high gel strength agar with excellent properties for tissue culture and microbiological work.
Molecular Biology Grade