Learn More
Alkali Scientific Agar, High Gel Strength, Powder, 500 G
SDP
Catalog No. 5000660634
Encompass_Preferred
Agar, High Gel Strength, Powder, 500 G is a smaller quantity of high-quality agar designed for preparing solid agar media in microbiological applications. This agar is known for its strong gel formation properties, making it ideal for laboratories that require firm and stable agar media for microbial culture work. It is used for preparing plates and slants that support bacterial, fungal, and yeast cultures. The 500 G package is perfect for smaller laboratories or those requiring smaller amounts of agar for occasional or lower-volume testing. The high gel strength ensures that the media maintains its integrity throughout incubation and microbial growth.
Catalog No. 50-006-60634 Supplier Alkali Scientific Supplier No. AGB012CP
Agar, High Gel Strength, Powder, 500 G is a smaller quantity of high-quality agar designed for preparing solid agar media in microbiological applications. This agar is known for its strong gel formation properties, making it ideal for laboratories that require firm and stable agar media for microbial culture work. It is used for preparing plates and slants that support bacterial, fungal, and yeast cultures. The 500 G package is perfect for smaller laboratories or those requiring smaller amounts of agar for occasional or lower-volume testing. The high gel strength ensures that the media maintains its integrity throughout incubation and microbial growth.