Learn More
Bioworld Agar, High Gel Strength, 1 kg
Catalog No. 50255206
Encompass_Preferred
AgarPurified high gel strength agar with excellent properties for tissue culture and microbiological workGel Strength: 900 g/cm2Working Concentration: 5.0 g/LSulfated ash: <6%This product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000
Catalog No. 50-255-206 Supplier Bioworld Supplier No. 401000642
AgarPurified high gel strength agar with excellent properties for tissue culture and microbiological workGel Strength: 900 g/cm2Working Concentration: 5.0 g/LSulfated ash: <6%This product is sold for laboratory research use only.HS Code : 130231000RTECS Number: AW7950000