Learn More
Medchemexpress LLC Agar, microbiology tested for cell culture | 9002-18-0 | MFCD00081288 | 336.3 g/mol | C14H24O9 | 100 G
SDP
Catalog No. 5000282858
Encompass_Preferred
Agar is a gel-forming polysaccharide extracted from red algae and supplied as a microbiology-tested powder for use as a solidifying agent in microbiological culture media, plant tissue culture, and related cell culture applications. It forms reversible physical gels and is provided with batch analytical data for laboratory use.
  • Microbiology tested for cell culture.
  • Powder form, off-white to light yellow.
  • Gel strength greater than 800 g/cm2.
  • Analytical profile: moisture ~15%, total ash ~3%, insoluble impurities ~0.6%.
  • Store at room temperature; typical shelf life about 3 years.
Catalog No. 50-002-82858 Supplier Medchemexpress LLC Supplier No. HYW134423100G
Agar is a gel-forming polysaccharide extracted from red algae and supplied as a microbiology-tested powder for use as a solidifying agent in microbiological culture media, plant tissue culture, and related cell culture applications. It forms reversible physical gels and is provided with batch analytical data for laboratory use.
  • Microbiology tested for cell culture.
  • Powder form, off-white to light yellow.
  • Gel strength greater than 800 g/cm2.
  • Analytical profile: moisture ~15%, total ash ~3%, insoluble impurities ~0.6%.
  • Store at room temperature; typical shelf life about 3 years.