Learn More
Medchemexpress LLC Agar, microbiology tested, for cell culture | 9002-18-0 | MFCD00081288 | 336.33 g/mol | C14H24O9 | 25 G
SDP
Catalog No. 5000282694
Encompass_Preferred
Agar, microbiology tested, for cell culture is a microbiology-grade agar composed of agarose and agar gel isolated from red algae. Supplied as an off-white to light-yellow powder, it forms a reversible gel via hydrogen bonding and is intended for research use. Typical applications include microorganism culture media, plant tissue culture, food additive research, and fingerprint recognition.
  • Microbiology tested for laboratory research use.
  • Off-white to light-yellow powder for easy handling and dissolution.
  • Forms a reversible gel via hydrogen bonding.
  • Suitable for microorganism culture media and plant tissue culture.
  • Stable at room temperature for extended storage.
  • Reported ash of 2.7% and moisture (loss on drying) of 9% per COA.
Catalog No. 50-002-82694 Supplier Medchemexpress LLC Supplier No. HYW13442325G
Agar, microbiology tested, for cell culture is a microbiology-grade agar composed of agarose and agar gel isolated from red algae. Supplied as an off-white to light-yellow powder, it forms a reversible gel via hydrogen bonding and is intended for research use. Typical applications include microorganism culture media, plant tissue culture, food additive research, and fingerprint recognition.
  • Microbiology tested for laboratory research use.
  • Off-white to light-yellow powder for easy handling and dissolution.
  • Forms a reversible gel via hydrogen bonding.
  • Suitable for microorganism culture media and plant tissue culture.
  • Stable at room temperature for extended storage.
  • Reported ash of 2.7% and moisture (loss on drying) of 9% per COA.