Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar microbiology tested s10KG
Catalog No. 502902862
Encompass
Agar is a hydrophilic colloid obtained from marine algae (Rhodophyceae). It is soluble in boiling water and is of significance to the microbiological area.
Catalog No. 50-290-2862 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A129610KG
Agar is a hydrophilic colloid obtained from marine algae (Rhodophyceae). It is soluble in boiling water and is of significance to the microbiological area.