Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar microbiology tested s1KG
Catalog No. 502901919
Encompass
Agar is a hydrophilic colloid obtained from marine algae (Rhodophyceae). It is soluble in boiling water and is of significance to the microbiological area.
Catalog No. 50-290-1919 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A12961KG
Agar is a hydrophilic colloid obtained from marine algae (Rhodophyceae). It is soluble in boiling water and is of significance to the microbiological area.